Learn More
American Research Products Inc Human Interleukin 10 IL 10 EL
Supplier:  American Research Products Inc CSBE04593HIS
Encompass
Human Interleukin 10 IL-10 ELISA Kit from CUSABIO Detection Range 3 9 pg/ml-250 pg/ml Sensitivity 0 975 pg/ml Sample serum plasma tissue homogenates cell culture supernates Method Sandwich
Catalog No. 50-003-26428