Learn More
Biomatik Corporation Human Interleukin 10 (IL10) CLIA Kit
Supplier:  Biomatik Corporation EKU08790
Encompass_Preferred
Human Interleukin 10 (IL10) CLIA Kit for detecting IL10 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.69-500pg/ml. Sensitivity: 0.29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P22301. Gene ID: 3586. Target Synonyms: CSIF; IL10A; TGIF; Cytokine Synthesis Inhibitory Factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8629