Learn More
Biomatik Corporation Human Interleukin 10 Receptor Beta (IL10Rb) ELISA Kit
Supplier:  Biomatik Corporation EKU08408
Encompass_Preferred
Human Interleukin 10 Receptor Beta (IL10Rb) ELISA Kit for detecting IL10Rb in tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 78-5,000pg/ml. Sensitivity: 29pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q08334. Gene ID: 3588. Target Synonyms: CDW210b; CD210-B; IL10R-B; IL10-RB; CRF2-4; CRFB4; IL-10R2; Interleukin-10 receptor subunit 2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5844