Learn More
Bioworld HUMAN INTERLEUKIN-11 (RECOM
Supplier:  Bioworld 220603751
Encompass_Preferred
HUMAN INTERLEUKIN-11 (RECOMBINANT): Recombinant Human Interleukin-11 (rHuIL-11) Source: E.Coli Description: Recombinant Human IL-11 produced in E.Coli is a single, non-glycosylated polypeptide chain containing 179 amino acids and hav...
Catalog No. 50-253-472