Learn More
Biomatik Corporation Human Interleukin 12 (IL12) CLIA Kit
Supplier:  Biomatik Corporation EKU08832
Encompass_Preferred
Human Interleukin 12 (IL12) CLIA Kit for detecting IL12 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 0.98pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P29459; P29460. Gene ID: 3592; 3593. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8617