Learn More
Biomatik Corporation Human Interleukin 12 Receptor Beta 1 (IL12Rb1) ELISA Kit
Supplier:  Biomatik Corporation EKU05194
Encompass_Preferred
Human Interleukin 12 Receptor Beta 1 (IL12Rb1) ELISA Kit for detecting IL12Rb1 in serum, plasma, tissue homogenates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P42701. Gene ID: 3594. Target Synonyms: CD212; IL12R-B1; IL12-RB1; IL-12R-Beta1; IL12RB. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8915