Learn More
Biomatik Corporation Human Interleukin 17B (IL17B) ELISA Kit
Supplier:  Biomatik Corporation EKU05241
Encompass_Preferred
Human Interleukin 17B (IL17B) ELISA Kit for detecting IL17B in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 5.5pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q9UHF5. Gene ID: 27190. Target Synonyms: IL-20; Cytokine Zcyto7; Neuronal interleukin-17-related factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6885