Learn More
Biomatik Corporation Human Interleukin 17B (IL17B) CLIA Kit
Supplier:  Biomatik Corporation EKU09697
Encompass_Preferred
Human Interleukin 17B (IL17B) CLIA Kit for detecting IL17B in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.52pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UHF5. Gene ID: 27190. Target Synonyms: IL-20; Cytokine Zcyto7; Neuronal interleukin-17-related factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5340