Learn More
Biomatik Corporation Human Interleukin 17C (IL17C) CLIA Kit
Supplier:  Biomatik Corporation EKU09551
Encompass_Preferred
Human Interleukin 17C (IL17C) CLIA Kit for detecting IL17C in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.64pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9P0M4. Gene ID: 27189. Target Synonyms: Cytokine CX2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8499