Learn More
American Research Products Inc Human interleukin 18 binding p
Catalog No. 5000328398
Encompass
Human interleukin-18 binding prorein (IL-18BP) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Sandwich
Catalog No. 50-003-28398 Supplier American Research Products Inc Supplier No. CSBE13687H
Human interleukin-18 binding prorein (IL-18BP) ELISA Kit from CUSABIO Detection Range 15 6 pg/ml-1000 pg/ml Sensitivity 3 9 pg/ml Sample serum plasma Method Sandwich