Learn More
Biomatik Corporation Human Interleukin 18 Binding Protein (IL18BP) CLIA Kit
Supplier:  Biomatik Corporation EKU09651
Encompass_Preferred
Human Interleukin 18 Binding Protein (IL18BP) CLIA Kit for detecting IL18BP in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.18pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: O95998. Gene ID: 10068. Target Synonyms: IL18BPa; MC51L-53L-54L Homolog Gene Product; Tadekinig-alfa. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5766