Learn More
Biomatik Corporation Human Interleukin 19 (IL19) CLIA Kit
Supplier:  Biomatik Corporation EKU09512
Encompass_Preferred
Human Interleukin 19 (IL19) CLIA Kit for detecting IL19 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.53pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9UHD0. Gene ID: 29949. Target Synonyms: MDA1; ZMDA1, IL-10C, NG.1; Melanoma Differentiation Associated Protein-Like Protein. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6193