Learn More
Cusabio Technology LLC Human Interleukin 1BIL1B ELISA
Supplier:  Cusabio Technology LLC CSBE08053HIS
Encompass_Preferred
This kit detects IL1B in human serum plasma tissue homogenates and cell culture supernates using a quantitative sandwich ELISA method With a sensitivity of 0 586 pg/ml and a detection range spanning 2 344 pg/ml to 150 pg/ml it enables precise measurement of interleukin-1B concentrations in diverse biological samples The assay requires 50-100 l of sample volume and features a streamlined protocol with an incubation time of 1-5 hours Absorbance is read at 450 nm ensuring reliable quantification of IL-1B levels for research applications Designed for flexibility and accuracy this kit provides researchers with a robust tool to analyze inflammatory biomarkers in clinical or experimental settings while maintaining consistent performance across sample types
Catalog No. 50-000-49759