Learn More
Biomatik Corporation Human Interleukin 2 Receptor Gamma (IL2Rg) ELISA Kit
Supplier:  Biomatik Corporation EKU05281
Encompass_Preferred
Human Interleukin 2 Receptor Gamma (IL2Rg) ELISA Kit for detecting IL2Rg in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.057ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P31785. Gene ID: 3561. Target Synonyms: CD132; IL2R-g; IL2-RG; CD132; IMD4; p64; SCIDX; SCIDX1; Cytokine Receptor Common Gamma Chain; Severe Combined Immunodeficiency. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6883