Learn More
Biomatik Corporation Human Interleukin 20 (IL20) CLIA Kit
Supplier:  Biomatik Corporation EKU09882
Encompass_Preferred
Human Interleukin 20 (IL20) CLIA Kit for detecting IL20 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.61pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q9NYY1. Gene ID: 50604. Target Synonyms: IL10D; IL10-D; ZCYTO10; Cytokine Zcyto10. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8409