Learn More
Biomatik Corporation Human Interleukin 22 Receptor (IL22R) ELISA Kit
Supplier:  Biomatik Corporation EKU09207
Encompass_Preferred
Human Interleukin 22 Receptor (IL22R) ELISA Kit for detecting IL22R in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.059ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q8N6P7. Gene ID: 58985. Target Synonyms: IL22RA1; CRF2-9; ZcytoR11; Interleukin 22 Receptor,Alpha 1; Cytokine receptor class-II member 9. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4987