Learn More
Biomatik Corporation Human Interleukin 24 (IL24) CLIA Kit
Supplier:  Biomatik Corporation EKU09879
Encompass_Preferred
Human Interleukin 24 (IL24) CLIA Kit for detecting IL24 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.57pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q13007. Gene ID: 11009. Target Synonyms: C49A; FISP; IL10B; MDA7; Mob-5; ST16; Mda-7; Melanoma Differentiation Association Protein 7; Suppression Of Tumorigenicity 16. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8410