Learn More
American Research Products Inc Human Interleukin 27 subunit b
Supplier:  American Research Products Inc CSBEL007360HU
Encompass
Human Interleukin-27 subunit beta (EBI3) ELISA kit from CUSABIO Detection Range 12 5 pg/mL-400 pg/mL Sensitivity 6 25 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28236