Learn More
Biomatik Corporation Human Interleukin 31 (IL31) CLIA Kit
Supplier:  Biomatik Corporation EKU09659
Encompass_Preferred
Human Interleukin 31 (IL31) CLIA Kit for detecting IL31 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.47pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: Q6EBC2. Gene ID: 386653. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6489