Learn More
Biomatik Corporation Human Interleukin 32 (IL32) CLIA Kit
Supplier:  Biomatik Corporation EKU09701
Encompass_Preferred
Human Interleukin 32 (IL32) CLIA Kit for detecting IL32 in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 1.37-1,000pg/ml. Sensitivity: 0.54pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P24001. Gene ID: 9235. Target Synonyms: NK4; TAIF; TAIFb; TAIFd; Natural Killer Cell Transcript 4; Natural killer cells protein 4; Tumor necrosis factor alpha-inducing factor. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6639