Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Human IL-4 Recombinant Protein

Catalog No. ENRIL4I
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ENRIL4I 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ENRIL4I Supplier Invitrogen™ Supplier No. RIL4I
Add to Cart
Edge
Add to Cart

Recombinant Protein

Mature human IL-4 was expressed in Pichia pastoris, X-33. GenBank Accession # P05112, amino acids 25 - 153. For protein verification, the expressed protein was N-terminal sequenced and begins with the following 10 amino acids: HKCDITLQEI. Purity is >95% by SDS-PAGE analyses. Activity: The biological activity of recombinant human IL-4 was determined by measuring the proliferation of the factor-dependent human T-helper cell line, TF-1. Use: For in vitro applications. A dose-response assay should be performed to determine the optimal concentration for specific experiments.

Interleukin-4 (IL-4) is a pleiotropic, immune-modulatory cytokine produced by activated T cells, and serves as a ligand for interleukin 4 receptor. IL-4 has been found to have the broadest range of actions. The interleukin 4 receptor also binds to IL13, which may contribute to many overlapping functions of this cytokine and IL13. STAT6, a signal transducer and activator of transcription, has been shown to play an important role in mediating the immune regulatory signal of IL-4. The IL-4 gene, IL-3, IL-5, IL-13, and CSF2 form a cytokine gene cluster on chromosome 5q. In particular, IL-4 is located in close proximity to IL13 on the chromosome. Further, the IL-4 gene, IL13 and IL5 are found to be coordinated by several long-range regulatory elements over a 120 kilobase range on the chromosome. IL-4 delta 2 is a known splice variant of IL-4.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number P05112
For Use With (Application) Control, ELISA, Immunoprecipitation, Western Blot
Formulation PBS with no preservative
Gene ID (Entrez) 3565
Molecular Weight (g/mol) 15 kDa
Name Human IL-4
Quantity 20 μg
Source Yeast
Gene Alias B cell growth factor 1; B_cell stimulatory factor 1; B-cell growth factor 1; B-cell IgG differentiation factor; B-cell stimulatory factor 1; BCGF1; BCGF-1; binetrakin; BSF1; BSF-1; cytokine; H-IL-4; IGG1 induction factor; IL2; IL4; Il-4; Il4e12; ILN; Interleukin; interleukin 4; interleukin 4 variant 2; interleukin 4 variant IL-4 delta2; interleukin 4 variant IL-4 delta3; interleukin 4 variant IL-4 int2A; interleukin 4 variant IL-4 int3A; interleukin 4 variant IL-4 int3B; Interleukin4; interleukin-4; interleukin-4 precursor; lymphocyte stimulatory factor 1; MGC79402; M-IL-4; Pitrakinra
Common Name IL-4
Gene Symbol IL4
Biological Activity Determined by the proliferation of human TF-1 cells.
Conjugate Unconjugated
Species Yeast
Recombinant Recombinant
Sequence Human IL-4, amino acids 25-153 (Accession # P05112)
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System P. pastoris
Form Liquid
Protein Subtype Recombinant
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.