Learn More
American Research Products Inc Human Interleukin 5 IL 5 ELIS
Catalog No. 5000326585
Encompass
Human Interleukin 5 IL-5 ELISA KIT from CUSABIO Detection Range 3 9 pg/ml-250 pg/ml Sensitivity 0 97 pg/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26585 Supplier American Research Products Inc Supplier No. CSBE04636H
Human Interleukin 5 IL-5 ELISA KIT from CUSABIO Detection Range 3 9 pg/ml-250 pg/ml Sensitivity 0 97 pg/ml Sample serum plasma tissue homogenates Method Sandwich