Learn More
Bioworld HUMAN INTERLEUKIN-6 (RECOMB
Supplier:  Bioworld 220603871
Encompass_Preferred
HUMAN INTERLEUKIN-6 (RECOMBINANT): Recombinant Human Interleukin-6 (IL-6 Cytokines of the IL6/GCSF/MGF family are glycoproteins of about 170 to 180 amino acid residues that contains four conserved cysteine residues involved in two disu...
Catalog No. 50-253-484