Learn More
Cusabio Technology LLC Human Interleukin 9IL-9 ELISA
Catalog No. 5000051784
Encompass_Preferred
This kit detects IL9 in human serum plasma cell culture supernates and tissue homogenates with high precision offering a broad detection range of 15 6 pg/mL to 1000 pg/mL and exceptional sensitivity down to 3 9 pg/mL Utilizing a quantitative sandwich ELISA methodology it delivers reliable results within 1-5 hours using only 50-100 L of sample per test The assay employs a 450 nm wavelength measurement to ensure accurate quantification of IL9 concentrations making it suitable for both research and clinical applications requiring precise monitoring of interleukin-9 levels across diverse biological matrices Its optimized protocol balances speed and accuracy for efficient performance in laboratory settings
Catalog No. 50-000-51784 Supplier Cusabio Technology LLC Supplier No. CSBE04642H
This kit detects IL9 in human serum plasma cell culture supernates and tissue homogenates with high precision offering a broad detection range of 15 6 pg/mL to 1000 pg/mL and exceptional sensitivity down to 3 9 pg/mL Utilizing a quantitative sandwich ELISA methodology it delivers reliable results within 1-5 hours using only 50-100 L of sample per test The assay employs a 450 nm wavelength measurement to ensure accurate quantification of IL9 concentrations making it suitable for both research and clinical applications requiring precise monitoring of interleukin-9 levels across diverse biological matrices Its optimized protocol balances speed and accuracy for efficient performance in laboratory settings