Learn More
American Research Products Inc Human Interleukin17C IL17C E
Catalog No. 5000328545
Encompass
Human Interleukin17C (IL17C) ELISA Kit from CUSABIO Detection Range 0 156 ng/ml -10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-28545 Supplier American Research Products Inc Supplier No. CSBE15917H
Human Interleukin17C (IL17C) ELISA Kit from CUSABIO Detection Range 0 156 ng/ml -10 ng/ml Sensitivity 0 039 ng/ml Sample serum plasma tissue homogenates Method Sandwich