Learn More
Biomatik Corporation Human Ionized Calcium-binding Adapter Molecule 1 (IBA1) CLIA Kit
Supplier:  Biomatik Corporation EKU09825
Encompass_Preferred
Human Ionized Calcium-binding Adapter Molecule 1 (IBA1) CLIA Kit for detecting IBA1 in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 2.74-2,000pg/ml. Sensitivity: 1.08pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Chemiluminescence. Assay Time: 2h, 40min. Accession Number: P55008. Gene ID: 199. Target Synonyms: AIF1; IRT1; Daintain; Protein G1; Allograft Inflammatory Factor 1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5079