Learn More
Abcam Human IP-10 ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB1731941X96TESTS
Encompass_Preferred
Human IP-10 ELISA Kit is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 2.6 pg/mL. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Colorimetric Sandwich ELISA - 450 nm readout : works on any standard plate reader - Cited in over 10 citations
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC1579514