Learn More
Abcam Human IP-10 ELISA Kit, Fluorescent, 1 x 96 Tests
Catalog No. 502711636
Encompass_Preferred
Human IP-10 ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 1.039 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-271-1636 Supplier Abcam Supplier No. AB3169031X96TESTS
Human IP-10 ELISA Kit, Fluorescent is a single-wash 90-min Simplestep used to quantify Human IP-10 with a sensitivity of 1.039 pg/ml. The assay uses a simple mix-wash-read protocol with just one incubation and one wash step. - Fluorescent Sandwich ELISA - 530/570/590 nm readout : works on any standard plate reader
The product is subject to the following: Abcam Restricted Use Statement