Learn More
FUJIFILM BIOSCIENCES INC Human IP-10 (CXCL10) , Recombinant Protein, Shenandoah Biotechnology, 25 ug vial
Supplier:  FUJIFILM BIOSCIENCES INC 10012725UG
Encompass_Preferred
Interferon gamma-induced protein 10 (IP-10), or CXCL10, is a chemokine secreted by monocytes, endothelial cells and fibroblasts in response to interferon gamma (IFN-ɣ). IP-10 functions as a chemoattractant for activated T cells, monocytes, dendritic, and natural killer (NK) cells that express the G protein-coupled receptor CXCR3. IP-10 is an important factor in autoimmune diseases such as Hashimoto's thyroiditis, Graves' disease, and Type 1 diabetes mellitus.
Catalog No. 50-990-647