Learn More
BIOHIPPO Human IPF Microsample
SDP
Catalog No. 5000313910
Encompass
Human IPF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IPF in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IPF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IPF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IPF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13910 Supplier BIOHIPPO Supplier No. BHE15210620
Human IPF Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IPF in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 051 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IPF Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IPF Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IPF biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color