Learn More
Abcam Human IRAK4 Antibody Pair - BSA and Azide free, 10 x 96 Tests
Catalog No. 502675655
Encompass_Preferred
Human IRAK4 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human IRAK4.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-5655 Supplier Abcam Supplier No. AB24185910X96TESTS
Human IRAK4 Antibody Pair - BSA and Azide free is a kit containing recombinant capture and detector antibodies in a carrier-free formulation for the measurement of Human IRAK4.
The product is subject to the following: Abcam Restricted Use Statement