Learn More
BIORTUS USA INC Human IRAK4 Protein, 100UG
Catalog No. NC2953315
Encompass_Preferred
Gene Name: IRAK4. Uniprot: Q9NWZ3. Construct: His-TEV-IRAK4(E154-S460 end). Expression Host: Insect cell. Purity: 90%.
Catalog No. NC2953315 Supplier BIORTUS USA INC Supplier No. BP0627701A
Gene Name: IRAK4. Uniprot: Q9NWZ3. Construct: His-TEV-IRAK4(E154-S460 end). Expression Host: Insect cell. Purity: 90%.