Learn More
BIOHIPPO Human IREM1 Microsample
SDP
Catalog No. 5000313840
Encompass
Human IREM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IREM1 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IREM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IREM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IREM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13840 Supplier BIOHIPPO Supplier No. BHE15211596
Human IREM1 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of IREM1 in Tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 16-10 ng/mL with a sensitivity of 0 064 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human IREM1 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human IREM1 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human IREM1 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color