Learn More
Sino Biological Anti-IRF6 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 105143T32100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human IRF6, and purified by antigen affinity chromatography.
Catalog No. 50-241-2271