Learn More
Biomatik Corporation Human Ischemia Modified Albumin (IMA) CLIA Kit
Supplier:  Biomatik Corporation EKU11934
Encompass_Preferred
Human Ischemia Modified Albumin (IMA) CLIA Kit for detecting IMA in serum, plasma, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 370.4-30,000ng/ml. Sensitivity: 131.3ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Chemiluminescence. Assay Time: 2h. Accession Number: P02768. Gene ID: 213. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5452