Learn More
Raybiotech Inc Human/Mouse/Rat Isoprostane EIA Kit.
Supplier:  Raybiotech Inc EIAIPF2A1
Encompass_Preferred
Optimized for serum and cell culture media. Competition-based ELISA on a 96-well strip plate.
Catalog No. 50-194-8822