Learn More
Sino Biological Recombinant Human CXCL11 Protein 100 µg
Catalog No. 502247018
Encompass_Preferred
A DNA sequence encoding the mature form of human CXCL11 (O14625) (Phe22-Phe94) was expressed with an initial Met.
Catalog No. 50-224-7018 Supplier Sino Biological Supplier No. 10876HNAE100
Add to Cart
Add to Cart
A DNA sequence encoding the mature form of human CXCL11 (O14625) (Phe22-Phe94) was expressed with an initial Met.