Learn More
Sino Biological Human Integrin alpha 2b beta 3 Heterodimer Protein (I874S, Flag & His Tag)
Supplier:  Sino Biological CT131H2508H100
Encompass_Preferred
A DNA sequence encoding the human ITGA2B (P08514-1)(Met1-Arg993, I874S) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human ITGB3 (P05106-1) (Met1-Asp718) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the Integrin alpha 2b beta 3 Heterodimer was purified.
Catalog No. 50-255-2589