Learn More
Sino Biological Recombinant Human Integrin alpha 4 beta 7 Protein (His Tag)
Catalog No. 502992825
Encompass_Preferred
A DNA sequence encoding the Human ITGA4 (P13612-1) (Tyr34-Thr977) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Human ITGB7 (P26010-1) (Glu20-His723) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the Human Integrin alpha 4 beta 7 Heterodimer was purified.
Catalog No. 50-299-2825 Supplier Sino Biological Supplier No. CT127H08H100
A DNA sequence encoding the Human ITGA4 (P13612-1) (Tyr34-Thr977) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the Human ITGB7 (P26010-1) (Glu20-His723) was expressed, constructed the plasmid 2. The two plasmids were co-expressed and the Human Integrin alpha 4 beta 7 Heterodimer was purified.