Learn More
Sino Biological Anti-Integrin alpha 5 beta 1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological CT014MM05P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITGA5 & ITGB1 Heterodimer (rh ITGA5 & ITGB1 Heterodimer; Catalog#CT014-H2508H; P08648 & P05556-1; Met1-Tyr995 & Met1-Asp728) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-0034