Learn More
Sino Biological Human ITGA5 & ITGB1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological CT014MM01100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITGA5 & ITGB1 Heterodimer (rh ITGA5 & ITGB1 Heterodimer; Catalog#CT014-H2508H; P08648, P05556-1; Met 1-Tyr 995, Met 1-Asp 728). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3010