Learn More
Sino Biological Anti-Integrin alpha 6 beta 1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological CT013RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human ITGA6 & ITGB1 Heterodimer (rh ITGA6 & ITGB1 Heterodimer; Catalog#CT013-H2508H; P23229-3 & P05556-1; Met 1-Ser 1011, Met 1-Asp 728). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-2624