Learn More
ACROBiosystems Human Integrin isoform alpha-7X2B beta 1 (ITGA7X2B&ITGB1) Heterodimer Protein, His Tag&Tag Free
Catalog No. 502331667
Encompass_Preferred
Human ITGA7X2B&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W8) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).
Catalog No. 50-233-1667 Supplier ACROBiosystems Supplier No. IT1H52W81MG
Human ITGA7X2B&ITGB1 Heterodimer Protein, His Tag&Tag Free (IT1-H52W8) is expressed from human 293 cells (HEK293). It contains AA Phe 34 - Trp 1038 (ITGA7X2B) & Gln 21 - Asp 728 (ITGB1) (Accession # Q13683-7 (ITGA7X2B) & P05556-1 (ITGB1)).