Learn More
BIOHIPPO Human ITGaM Microsample
SDP
Catalog No. 5000315863
Encompass
Human ITGaM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ITGaM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ITGaM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ITGaM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ITGaM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15863 Supplier BIOHIPPO Supplier No. BHE15211484
Human ITGaM Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of ITGaM in Serum plasma tissue homogenates and other biological fluids It provides a detection range of 31 25-2000 pg/mL with a sensitivity of 13 1 pg/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human ITGaM Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human ITGaM Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human ITGaM biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color