Learn More
Sino Biological Human ITGAV & ITGB6 MAb 100ul (Sino Biological)
Supplier:  Sino Biological CT039MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human Human ITGAV & ITGB6 Heterodimer (rh Human ITGAV & ITGB6 Heterodimer; Catalog#CT039-H2508H; P06756-1 & P18564-1; Met1-Val992 & Met1-Asn707). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3402