Learn More
Sino Biological Recombinant Human Integrin alpha V beta 8 Protein (His Tag), HPLC-verified 1
Supplier:  Sino Biological CT099H08H1
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Val992) of human ITGAV (P06756) was fused with a C-terminal polyhistidine tag was expressed., constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Arg684) of human ITGB8 (P26012) constructed the plasmid 2. The two plasmids were co-expressed and the human ITGAV&ITGB8 heterodimer was purified.
Catalog No. 50-224-6672