Learn More
Sino Biological Recombinant Human Integrin alpha V beta 1 Protein (Flag & His Tag)
Supplier:  Sino Biological CT130H2508H1100
Encompass_Preferred
A DNA sequence encoding the human ITGAV (P06756-1) (Met1-Val992) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human ITGB1 (P05556-1) (Met1-Asp728) was fused with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the human ITGAV & ITGB1 heterodimer was purified.
Catalog No. 50-255-2591