Learn More
Sino Biological Recombinant Human Integrin alpha V beta 5 Protein (Flag & His Tag), HPLC-verified
Supplier:  Sino Biological CT129H2508H1
Encompass_Preferred
A DNA sequence encoding the human ITGAV (P06756-1)(Met1-Val992) was fused with a flag tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the human ITGB5 (P18084-1) (Met1-Asn719) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 2. The two plasmids were co-expressed and the Integrin alpha V beta 5 Heterodimer was purified.
Catalog No. 50-255-2594