Learn More
Sino Biological Recombinant Human Integrin alpha X beta 2 Protein (His Tag)
Supplier:  Sino Biological CT017H08H150
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met1-Pro1107) of human ITGAX (P20702) was expressed with a polyhistidine tag at the C-terminus, constructed the plasmid 1; A DNA sequence encoding the extracellular domain (Met1-Asn700) of human ITGB2 (P05107), constructed the plasmid 2. The two plasmids were co-expressed and the heterodimer was purified.
Catalog No. 50-255-2623