Learn More
Sino Biological Human ITGB1 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10587MM06100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human ITGB1 / Integrin beta-1 / CD29 (rh ITGB1 / Integrin beta-1 / CD29; Catalog#10587-H08H1; CAA30790.1; Met1-Asp728). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3628